ENAH3035CB71 Enamine

5-(2-hydroxyphenyl)-1,2-oxazole-3-carboxylic acid

  • MDL number MFCD05237206
  • Cas Number : 33282-12-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-152805

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-152805
Availability details for
Quantity
  • Availability
  • Add to Cart