ENAH3035CB82 Enamine

4-(propan-2-yl)oxane-2,6-dione

  • MDL number MFCD28362356
  • Cas Number : 4160-81-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-155524

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155524
Availability details for
Quantity
  • Availability
  • Add to Cart