ENAH3035CB85 Enamine

4-(bromomethyl)-1-(difluoromethoxy)-2-methoxybenzene

  • MDL number MFCD11180531
  • Cas Number : 853771-74-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-155655

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155655
Availability details for
Quantity
  • Availability
  • Add to Cart