ENAH3035CB86 Enamine

4-iodo-3-methyl-1,2-thiazol-5-amine

  • MDL number MFCD20257781
  • Cas Number : 851049-22-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-155809

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-155809
Availability details for
Quantity
  • Availability
  • Add to Cart