ENAH3035CB8D Enamine

4-[1-(3-chlorophenyl)-1H-1,2,3-triazole-4-carbonyl]thiomorpholine

  • MDL number MFCD15129559
  • Cas Number : 951612-19-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-157186

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-157186
Availability details for
Quantity
  • Availability
  • Add to Cart