ENAH3035CB99 Enamine

2-methoxyethyl 5-methyl-4-oxo-2-(pyridin-3-yl)-3H,4H-thieno[2,3-d]pyrimidine-6-carboxylate

  • Cas Number : 727982-66-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-16547

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-16547
Availability details for
Quantity
  • Availability
  • Add to Cart