ENAH3035CBAD Enamine

4-chloro-2-fluoro-N-methoxy-N-methylbenzamide

  • MDL number MFCD03428601
  • Cas Number : 198967-23-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-171505

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-171505
Availability details for
Quantity
  • Availability
  • Add to Cart