ENAH3035CBE4 Enamine

4-(propan-2-yl)-5-(trifluoromethyl)-1H-pyrazole-3-carboxylic acid

  • MDL number MFCD31719560
  • Cas Number : 2103589-03-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-184868

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-184868
Availability details for
Quantity
  • Availability
  • Add to Cart