ENAH3035CBE6 Enamine

3,3-dimethyloxetane-2-carboxylic acid

  • MDL number MFCD04973502
  • Cas Number : 20967-49-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-185317

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-185317
Availability details for
Quantity
  • Availability
  • Add to Cart