ENAH3035CBF6 Enamine

2-amino-3,6-dimethoxybenzoic acid

  • MDL number MFCD01926620
  • Cas Number : 50472-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-188452

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-188452
Availability details for
Quantity
  • Availability
  • Add to Cart