ENAH3035CBF9 Enamine

2-butylphenol

  • MDL number MFCD00102127
  • Molecular Weight : 150.22

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-189786

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-189786
Availability details for
Quantity
  • Availability
  • Add to Cart