ENAH3035CC11 Enamine

methyl 4-cyanopyridine-2-carboxylate

  • MDL number MFCD14582015
  • Cas Number : 142729-98-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-195174

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195174
Availability details for
Quantity
  • Availability
  • Add to Cart