ENAH3035CC13 Enamine

2-(2-oxoazepan-4-yl)acetic acid

  • MDL number MFCD20647118
  • Cas Number : 10139-43-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-195590

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-195590
Availability details for
Quantity
  • Availability
  • Add to Cart