ENAH3035CC18 Enamine

cyclopropylmethyl methanesulfonate

  • MDL number MFCD25966289
  • Cas Number : 696-77-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-196659

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-196659
Availability details for
Quantity
  • Availability
  • Add to Cart