ENAH3035CC1A Enamine

2-phenylcyclohexan-1-amine hydrochloride

  • MDL number MFCD27953540
  • Cas Number : 22720-50-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-197108

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-197108
Availability details for
Quantity
  • Availability
  • Add to Cart