ENAH3035CC20 Enamine

tert-butyl N-(3-fluoro-4-nitrophenyl)carbamate

  • MDL number MFCD09953070
  • Cas Number : 658700-15-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-199082

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-199082
Availability details for
Quantity
  • Availability
  • Add to Cart