ENAH3035CC24 Enamine

2,3,5,6-tetrachloroaniline

  • MDL number MFCD00007659
  • Cas Number : 3481-20-7

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 106-108
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-20148

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-20148
Availability details for
Quantity
  • Availability
  • Add to Cart