ENAH3035CC4D Enamine

3-(4-nitrophenyl)-1H,7H-imidazo[1,5-a]pyrazin-1-one

  • Cas Number : 2253640-87-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-57804

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-57804
Availability details for
Quantity
  • Availability
  • Add to Cart