ENAH3035CC56 Enamine

ethyl 3-benzamido-2-oxo-6-(trifluoromethyl)-2H-pyran-5-carboxylate

  • MDL number MFCD01936259
  • Cas Number : 400081-64-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-62330

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-62330
Availability details for
Quantity
  • Availability
  • Add to Cart