ENAH3035CC67 Enamine

1-benzylazepan-3-amine

  • MDL number MFCD22057300
  • Cas Number : 134907-68-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-70548

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-70548
Availability details for
Quantity
  • Availability
  • Add to Cart