ENAH3035CC74 Enamine

methyl 2-amino-4-tert-butylthiophene-3-carboxylate

  • MDL number MFCD07368947
  • Cas Number : 349662-74-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 56-58
Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-75785

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-75785
Availability details for
Quantity
  • Availability
  • Add to Cart