ENAH3035CC8A Enamine

5-chloro-2-cyclopropylpyrimidine-4-carboxylic acid

  • MDL number MFCD16988026
  • Cas Number : 1240597-65-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-84736

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-84736
Availability details for
Quantity
  • Availability
  • Add to Cart