ENAH3035CC96 Enamine

3-(chloromethyl)-6-methoxy-2-methylpyridine

  • MDL number MFCD16610684
  • Cas Number : 853569-72-9

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-90415

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90415
Availability details for
Quantity
  • Availability
  • Add to Cart