ENAH3035CC97 Enamine

(6-methoxy-2-methylpyridin-3-yl)methanol

  • MDL number MFCD16611534
  • Cas Number : 32383-10-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-90416

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-90416
Availability details for
Quantity
  • Availability
  • Add to Cart