ENAH3035CCA3 Enamine

5-chloro-3-methyl-4-nitro-1-(propan-2-yl)-1H-pyrazole

  • MDL number MFCD21667694
  • Cas Number : 13551-75-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-95067

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-95067
Availability details for
Quantity
  • Availability
  • Add to Cart