ENAH3035CCAB Enamine

1,1,6-trimethyl-1,2,3,4-tetrahydronaphthalene

  • MDL number MFCD00049170
  • Cas Number : 475-03-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form liquid
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-96891

Safety Information

signalword Warning
pcodes P261,P264,P271,P273,P280,P302+P352,P304+P340,P305+P351+P338,P312,P332+P313,P337+P313,P362,P391,P403+P233,P405,P501
MSDS https://www.enaminestore.com/catalog/EN300-96891
pictograms GHS07,GHS09
hstatements H315,H319,H335,H411
Availability details for
Quantity
  • Availability
  • Add to Cart