ENAH3035CCC7 Enamine

5-(1,3-benzothiazol-2-yl)pyridin-3-amine

  • MDL number MFCD11867983
  • Cas Number : 1118624-94-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1013635

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1013635
Availability details for
Quantity
  • Availability
  • Add to Cart