ENAH3035CF21 Enamine

2-bromo-5-chloropyridine-3-carbaldehyde

  • MDL number MFCD16606499
  • Cas Number : 1227605-52-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. 4
Shipping Temperature Normal
Country of Origin Ukraine
Product Link https://www.enaminestore.com/catalog/EN300-201166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201166
Availability details for
Quantity
  • Availability
  • Add to Cart