ENAH3035CF24 Enamine

1,2,3,4,5,6-hexahydro-1-benzazocine-2,6-dione

  • MDL number MFCD00227772
  • Cas Number : 35402-59-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-201489

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-201489
Availability details for
Quantity
  • Availability
  • Add to Cart