ENAH3035CF29 Enamine

2-(1H-1,2,3-triazol-1-yl)ethane-1-sulfonamide

  • MDL number MFCD31719010
  • Cas Number : 1936564-20-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-202469

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202469
Availability details for
Quantity
  • Availability
  • Add to Cart