ENAH3035CF2A Enamine

ethyl 2-[benzyl(2-oxopropyl)amino]acetate

  • MDL number MFCD18431656
  • Cas Number : 15057-40-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-202675

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-202675
Availability details for
Quantity
  • Availability
  • Add to Cart