ENAH3035CF46 Enamine

3-(pyridin-4-yl)-1H-pyrazol-5-ol

  • MDL number MFCD02933185
  • Cas Number : 78584-36-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-208083

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208083
Availability details for
Quantity
  • Availability
  • Add to Cart