ENAH3035CF48 Enamine

2-(5-oxopyrrolidin-2-yl)propanedioic acid

  • MDL number MFCD31655100
  • Cas Number : 1934442-79-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-208453

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-208453
Availability details for
Quantity
  • Availability
  • Add to Cart