ENAH3035CF64 Enamine

tert-butyl N-(1-methyl-1H-indol-3-yl)carbamate

  • MDL number MFCD24467076
  • Cas Number : 933800-38-5
  • Molecular Weight : 246.31

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-215969

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-215969
Availability details for
Quantity
  • Availability
  • Add to Cart