ENAH3035CF67 Enamine

4-chloro-1-(propan-2-yl)piperidine hydrochloride

  • MDL number MFCD31719361
  • Cas Number : 5570-79-6

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 237-238
Purity 95
Salt Data HCl
Product Link https://www.enaminestore.com/catalog/EN300-217143

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-217143
Availability details for
Quantity
  • Availability
  • Add to Cart