ENAH3035CF7B Enamine

1-methyl-1H-1,2,3-benzotriazole-4-carboxylic acid

  • MDL number MFCD20661146
  • Cas Number : 1312556-53-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-219923

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-219923
Availability details for
Quantity
  • Availability
  • Add to Cart