ENAH3035CF7E Enamine

1-methyl-2-(trifluoromethyl)cyclopropane-1-carboxylic acid

  • MDL number MFCD31719030
  • Cas Number : 1934574-81-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-221166

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221166
Availability details for
Quantity
  • Availability
  • Add to Cart