ENAH3035CF80 Enamine

3a,4,5,6,7,7a-hexahydro-1,3-benzothiazol-2-amine

  • MDL number MFCD07174355
  • Cas Number : 84147-08-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-221289

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-221289
Availability details for
Quantity
  • Availability
  • Add to Cart