ENAH3035CF8A Enamine

4-bromo-2-(methoxymethoxy)benzene-1-sulfonyl chloride

  • MDL number MFCD31736437
  • Cas Number : 2092808-82-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-223435

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223435
Availability details for
Quantity
  • Availability
  • Add to Cart