ENAH3035CF8D Enamine

5-bromo-2-fluoro-4-hydroxybenzoic acid

  • MDL number MFCD27995580
  • Cas Number : 1781790-71-3

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-223978

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-223978
Availability details for
Quantity
  • Availability
  • Add to Cart