ENAH3035CF9A Enamine

2,2,2-trichloro-1-(5-methyl-1H-pyrrol-2-yl)ethan-1-one

  • MDL number MFCD04971439
  • Cas Number : 183268-72-6
  • Molecular Weight : 226.49

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Melting Point 104-106
Purity 95
Storage temp. RT
Physical Form powder
Shipping Temperature Normal
Country of Origin UA
Product Link https://www.enaminestore.com/catalog/EN300-243494

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-243494
Availability details for
Quantity
  • Availability
  • Add to Cart