ENAH3035CF9C Enamine

{7-oxabicyclo[2.2.1]heptan-2-yl}methanesulfonyl chloride

  • MDL number MFCD31719589
  • Cas Number : 1851602-43-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-243906

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-243906
Availability details for
Quantity
  • Availability
  • Add to Cart