ENAH3035CFA3 Enamine

3-(3-bromo-2-fluorophenyl)propanoic acid

  • MDL number MFCD18391365
  • Cas Number : 1261814-91-8

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-244603

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-244603
Availability details for
Quantity
  • Availability
  • Add to Cart