ENAH3035CFA6 Enamine

ethyl 3-(trifluoromethyl)-1,2,4-oxadiazole-5-carboxylate

  • MDL number MFCD11975643
  • Cas Number : 1245643-72-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-244921

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-244921
Availability details for
Quantity
  • Availability
  • Add to Cart