ENAH3035CFAA Enamine

2lambda6-thia-1-azabicyclo[3.1.0]hexane-2,2-dione

  • MDL number MFCD31719031
  • Cas Number : 291514-09-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-246532

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-246532
Availability details for
Quantity
  • Availability
  • Add to Cart