ENAH3035CFB6 Enamine

2-[(tert-butoxy)carbonyl]-2-azaspiro[3.3]heptane-1-carboxylic acid

  • MDL number MFCD31718781
  • Cas Number : 1934376-95-0

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-248979

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-248979
Availability details for
Quantity
  • Availability
  • Add to Cart