ENAH3035CFBC Enamine

7-fluoro-1,2,3,4-tetrahydroquinoline-6-carboxylic acid

  • MDL number MFCD31719035
  • Cas Number : 1936263-26-1

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-250055

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-250055
Availability details for
Quantity
  • Availability
  • Add to Cart