ENAH3035CFC3 Enamine

2-chloro-5-iodo-1,3-thiazole

  • MDL number MFCD21728909
  • Cas Number : 1518622-46-2

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-250972

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-250972
Availability details for
Quantity
  • Availability
  • Add to Cart