ENAH3035CFC5 Enamine

3-bromocyclobutane-1-carboxylic acid

  • MDL number MFCD20631705
  • Cas Number : 1378752-12-5

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-251359

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-251359
Availability details for
Quantity
  • Availability
  • Add to Cart