ENAH3035CFE3 Enamine

2-(1-{[(tert-butoxy)carbonyl]amino}ethyl)-4-methyl-1,3-thiazole-5-carboxylic acid

  • MDL number MFCD31719219
  • Cas Number : 2248382-93-4

Pricing & Availability

  Product ID Vendor SKU Availability Price () Quantity  

Description

Purity 95
Product Link https://www.enaminestore.com/catalog/EN300-1071467

Safety Information

MSDS https://www.enaminestore.com/catalog/EN300-1071467
Availability details for
Quantity
  • Availability
  • Add to Cart